ENA497679869 Enamine

4-bromo-7-fluoro-1,2-dihydroisoquinolin-1-one

  • MDL number MFCD15832831
  • Cas Number : 1207448-47-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-262666

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-262666
Availability details for
Quantity
  • Availability
  • Add to Cart