ENA497679871 Enamine

4-bromopyridine-2,5-diamine

  • MDL number MFCD28737808
  • Cas Number : 1806851-06-8
  • Molecular Weight : 188.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-bromopyridine-2,5-diamine
Inchi Code 1S/C5H6BrN3/c6-3-1-5(8)9-2-4(3)7/h1-2H,7H2,(H2,8,9)
Inchi Key SWLIOAHOELOSMF-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-262671

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-262671
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart