ENA497679874 Enamine

tert-butyl 5-oxo-1-(piperidin-4-yl)pyrrolidine-3-carboxylate hydrochloride

  • MDL number MFCD29763330
  • Cas Number : 1909306-10-0
  • Molecular Weight : 304.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 5-oxo-1-(piperidin-4-yl)pyrrolidine-3-carboxylate hydrochloride
Inchi Code 1S/C14H24N2O3.ClH/c1-14(2,3)19-13(18)10-8-12(17)16(9-10)11-4-6-15-7-5-11;/h10-11,15H,4-9H2,1-3H3;1H
Inchi Key RAJVWPXKXXZERA-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-262697

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-262697
Availability details for
Quantity
  • Availability
  • Add to Cart