ENA497679879 Enamine

2,5-dichloro-4-ethoxypyrimidine

  • MDL number MFCD23161540
  • Molecular Weight : 193.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,5-dichloro-4-ethoxypyrimidine
Inchi Code 1S/C6H6Cl2N2O/c1-2-11-5-4(7)3-9-6(8)10-5/h3H,2H2,1H3
Inchi Key USUPRXXHCFGENX-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-262702

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-262702
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart