ENA497679882 Enamine

3-[(3-fluorophenyl)methyl]-5H,6H,7H,8H-[1,2,4]triazolo[4,3-a]pyrazine

  • MDL number MFCD11856681
  • Cas Number : 1159553-28-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-262706

Safety Information

signalword Warning
pcodes P264,P270,P280,P301+P312,P302+P352,P330,P332+P313,P362,P501
hcodes Xi
MSDS https://www.enaminestore.com/catalog/EN300-262706
pictograms GHS07
hstatements H302,H315
Availability details for
Quantity
  • Availability
  • Add to Cart