ENA497679891 Enamine

methyl 6-(methylsulfanyl)pyridine-2-carboxylate

  • MDL number MFCD01928254
  • Cas Number : 74470-41-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-262725

Safety Information

signalword Warning
pcodes P264,P270,P280,P301+P312,P302+P352,P312,P330,P363,P501
hcodes Xn
MSDS https://www.enaminestore.com/catalog/EN300-262725
pictograms GHS07
hstatements H302,H312
Availability details for
Quantity
  • Availability
  • Add to Cart