ENA497679918 Enamine

methyl 5-amino-3-(2-hydroxyethoxy)-1-methyl-1H-pyrazole-4-carboxylate

  • MDL number MFCD24633941
  • Cas Number : 126865-28-9
  • Molecular Weight : 215.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 5-amino-3-(2-hydroxyethoxy)-1-methyl-1H-pyrazole-4-carboxylate
Inchi Code 1S/C8H13N3O4/c1-11-6(9)5(8(13)14-2)7(10-11)15-4-3-12/h12H,3-4,9H2,1-2H3
Inchi Key NUTVPCQGMFYYQY-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-262764

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-262764
Availability details for
Quantity
  • Availability
  • Add to Cart