ENA497679920 Enamine

1-(piperidin-4-yl)cyclopropan-1-ol

  • MDL number MFCD26938039
  • Cas Number : 1358782-67-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-262767

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-262767
Availability details for
Quantity
  • Availability
  • Add to Cart