ENA497679926 Enamine

5-carbamoyl-2,3-dimethylbenzene-1-sulfonyl chloride

  • MDL number MFCD21224333
  • Cas Number : 1462994-43-9
  • Molecular Weight : 247.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-262777

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-262777
Availability details for
Quantity
  • Availability
  • Add to Cart