ENA497679960 Enamine

N-[(4-hydroxy-3-methoxyphenyl)methyl]acetamide

  • MDL number MFCD18970641
  • Cas Number : 53527-04-1
  • Molecular Weight : 195.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-262931

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-262931
Availability details for
Quantity
  • Availability
  • Add to Cart