ENA497679977 Enamine

(8-fluoro-1,2,3,4-tetrahydroquinolin-3-yl)methanol

  • MDL number MFCD09895662
  • Cas Number : 1017191-02-4
  • Molecular Weight : 181.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-262991

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-262991
Availability details for
Quantity
  • Availability
  • Add to Cart