ENA497679987 Enamine

2-(bromomethyl)-6-methoxy-1,3-benzothiazole

  • MDL number MFCD11217503
  • Cas Number : 110704-18-2
  • Molecular Weight : 258.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-263009

Safety Information

signalword Warning
pcodes P264,P270,P280,P301+P312,P302+P352,P312,P330,P363,P501
MSDS https://www.enaminestore.com/catalog/EN300-263009
pictograms GHS07
hstatements H302,H312
Availability details for
Quantity
  • Availability
  • Add to Cart