ENA497679990 Enamine

4-(dibromomethyl)-3-nitrobenzonitrile

  • MDL number MFCD29763357
  • Cas Number : 122947-72-2
  • Molecular Weight : 319.94

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-263026

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-263026
Availability details for
Quantity
  • Availability
  • Add to Cart