ENA497680009 Enamine

methyl 4-acetamidobenzoate

  • MDL number MFCD00463106
  • Cas Number : 17012-22-5
  • Molecular Weight : 193.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-263089

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-263089
Availability details for
Quantity
  • Availability
  • Add to Cart