ENA497680018 Enamine

methyl 2-(dimethylamino)-3-hydroxybutanoate

  • MDL number MFCD29763365
  • Molecular Weight : 161.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-263124

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-263124
Availability details for
Quantity
  • Availability
  • Add to Cart