ENA497680034 Enamine

1-(3-chloro-4-methylphenyl)-2,3-dihydro-1H-imidazol-2-one

  • MDL number MFCD23697189
  • Cas Number : 1535310-94-1
  • Molecular Weight : 208.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-263158

Safety Information

signalword Warning
pcodes P264,P270,P280,P301+P312,P302+P352,P312,P330,P363,P501
MSDS https://www.enaminestore.com/catalog/EN300-263158
pictograms GHS07
hstatements H302,H312
Availability details for
Quantity
  • Availability
  • Add to Cart