ENA497680042 Enamine

5-methanesulfonylfuran-3-carboxylic acid

  • MDL number MFCD29763372
  • Cas Number : 1520606-32-9
  • Molecular Weight : 190.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-263180

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-263180
Availability details for
Quantity
  • Availability
  • Add to Cart