ENA497680043 Enamine

1,1,1-trifluoro-N-(2-hydroxyethyl)methanesulfonamide

  • MDL number MFCD12185396
  • Cas Number : 40630-40-8
  • Molecular Weight : 193.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-263182

Safety Information

signalword Danger
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P310,P312,P332+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-263182
pictograms GHS05,GHS07
hstatements H315,H318,H335
Availability details for
Quantity
  • Availability
  • Add to Cart