ENA497680047 Enamine

(2S)-2-[(4-fluorophenyl)formamido]-3-hydroxypropanoic acid

  • MDL number MFCD12795637
  • Cas Number : 1102888-26-5
  • Molecular Weight : 227.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-263186

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-263186
Availability details for
Quantity
  • Availability
  • Add to Cart