ENA497680050 Enamine

[(4-fluorophenyl)(1H-imidazol-2-yl)methyl](methyl)amine

  • MDL number MFCD23866801
  • Cas Number : 1518527-67-7
  • Molecular Weight : 205.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-263192

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-263192
Availability details for
Quantity
  • Availability
  • Add to Cart