ENA497680051 Enamine

4-(5-chloro-1-benzofuran-2-yl)piperidine

  • MDL number MFCD26672914
  • Cas Number : 54403-14-4
  • Molecular Weight : 235.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-263206

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-263206
Availability details for
Quantity
  • Availability
  • Add to Cart