ENA497680056 Enamine

2-ethenyl-6-methylpyrazine

  • MDL number MFCD22200211
  • Cas Number : 13925-09-2
  • Molecular Weight : 120.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-263260

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-263260
Availability details for
Quantity
  • Availability
  • Add to Cart