ENA497680058 Enamine

tert-butyl 4-(3-oxopropyl)piperidine-1-carboxylate

  • MDL number MFCD09910237
  • Cas Number : 165528-85-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-263262

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-263262
Availability details for
Quantity
  • Availability
  • Add to Cart