ENA497680071 Enamine

3-amino-6,6-dimethylcyclohex-2-en-1-one

  • MDL number MFCD19203859
  • Cas Number : 24068-46-0
  • Molecular Weight : 139.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-263277

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-263277
Availability details for
Quantity
  • Availability
  • Add to Cart