ENA497680076 Enamine

1-(2-methylphenyl)cyclobutan-1-amine hydrochloride

  • MDL number MFCD09864762
  • Cas Number : 1228880-11-2
  • Molecular Weight : 197.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-263286

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-263286
Availability details for
Quantity
  • Availability
  • Add to Cart