ENA497680082 Enamine

3-(2-aminoethyl)-2,3-dihydro-1,3-thiazol-2-one

  • MDL number MFCD18328117
  • Cas Number : 1340039-62-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-263300

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-263300
Availability details for
Quantity
  • Availability
  • Add to Cart