ENA497680090 Enamine

methyl N-ethenylcarbamate

  • MDL number MFCD19216881
  • Cas Number : 15204-93-0
  • Molecular Weight : 101.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-263315

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-263315
Availability details for
Quantity
  • Availability
  • Add to Cart