ENA497680107 Enamine

6-fluoro-2-[(piperidin-4-yl)methyl]-1,3-benzoxazole

  • MDL number MFCD24883117
  • Cas Number : 1521612-18-9
  • Molecular Weight : 234.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-263343

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-263343
Availability details for
Quantity
  • Availability
  • Add to Cart