ENA497680115 Enamine

1-(4-fluorophenyl)-5-methyl-1H-pyrazol-4-amine hydrochloride

  • MDL number MFCD29763389
  • Cas Number : 1909316-81-9
  • Molecular Weight : 227.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-263365

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-263365
Availability details for
Quantity
  • Availability
  • Add to Cart