ENA497680127 Enamine

4-[3-(4-methylpiperazin-1-yl)propoxy]aniline

  • MDL number MFCD11551843
  • Cas Number : 220822-26-4
  • Molecular Weight : 249.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-263397

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-263397
Availability details for
Quantity
  • Availability
  • Add to Cart