ENA497680236 Enamine

6-bromo-8-(trifluoromethyl)-3,4-dihydro-2H-1,4-benzoxazin-3-one

  • MDL number MFCD29907075
  • Cas Number : 1909311-73-4
  • Molecular Weight : 296.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-263532

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-263532
Availability details for
Quantity
  • Availability
  • Add to Cart