ENA497680244 Enamine

tert-butyl (1r,4r)-4-(hydrazinecarbonyl)cyclohexane-1-carboxylate

  • MDL number MFCD29907077
  • Cas Number : 1909294-66-1
  • Molecular Weight : 242.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-263637

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-263637
Availability details for
Quantity
  • Availability
  • Add to Cart