ENA497680261 Enamine

1-(1-methyl-1H-pyrazol-4-yl)-1H-pyrrole-3-carbaldehyde

  • MDL number MFCD29907079
  • Cas Number : 1909320-16-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-263843

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-263843
Availability details for
Quantity
  • Availability
  • Add to Cart