ENA497680264 Enamine

3-(4-nitrophenyl)-1,2-oxazole-5-sulfonyl chloride

  • MDL number MFCD29763435
  • Cas Number : 1909337-76-3
  • Molecular Weight : 288.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-263894

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-263894
Availability details for
Quantity
  • Availability
  • Add to Cart