ENA497680274 Enamine

9-hydroxy-4lambda6-thia-5-azatricyclo[4.2.1.0,3,7]nonane-4,4-dione

  • MDL number MFCD29907085
  • Cas Number : 1453422-57-5
  • Molecular Weight : 189.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-264041

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-264041
Availability details for
Quantity
  • Availability
  • Add to Cart