ENA497680297 Enamine

4-{[3-(2-methylcyclopropyl)-1,2,4-oxadiazol-5-yl]methyl}piperidine hydrochloride

  • MDL number MFCD29907112
  • Cas Number : 1909318-66-6
  • Molecular Weight : 257.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-264756

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-264756
Availability details for
Quantity
  • Availability
  • Add to Cart