ENA497680300 Enamine

1-(4-nitro-2,3-dihydro-1H-indol-1-yl)ethan-1-one

  • MDL number MFCD28680430
  • Cas Number : 84807-25-0
  • Molecular Weight : 206.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-263670

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-263670
Availability details for
Quantity
  • Availability
  • Add to Cart