ENA497680302 Enamine

1-benzyl-1-nitrosourea

  • MDL number MFCD01676879
  • Cas Number : 775-11-1
  • Molecular Weight : 179.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 100-101
Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-263673

Safety Information

signalword Danger
pcodes P201,P202,P264,P270,P281,P301+P310,P308+P313,P330,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-263673
pictograms GHS06,GHS08
hstatements H301,H350
Availability details for
Quantity
  • Availability
  • Add to Cart