ENA497680316 Enamine

3-(3-bromothiophen-2-yl)-3-oxopropanenitrile

  • MDL number MFCD18888240
  • Cas Number : 1339722-54-1
  • Molecular Weight : 230.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-263722

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-263722
Availability details for
Quantity
  • Availability
  • Add to Cart