ENA497680328 Enamine

tert-butyl N-[(1S)-1-carbamoyl-2-phenylethyl]carbamate

  • MDL number MFCD00272775
  • Cas Number : 35150-06-2
  • Molecular Weight : 264.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-263517

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-263517
Availability details for
Quantity
  • Availability
  • Add to Cart