ENA497680336 Enamine

2-(3-bromopropyl)-1,3-difluorobenzene

  • MDL number MFCD09744357
  • Cas Number : 401939-94-4
  • Molecular Weight : 235.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-263847

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-263847
Availability details for
Quantity
  • Availability
  • Add to Cart