ENA497680337 Enamine

4-(2,6-difluorophenyl)butanenitrile

  • MDL number MFCD09744527
  • Cas Number : 1057672-27-1
  • Molecular Weight : 181.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-263848

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-263848
Availability details for
Quantity
  • Availability
  • Add to Cart