ENA497680345 Enamine

2-cyclobutylpiperidine hydrochloride

  • MDL number MFCD28895164
  • Cas Number : 1889294-03-4
  • Molecular Weight : 175.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-263481

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-263481
Availability details for
Quantity
  • Availability
  • Add to Cart