ENA497680353 Enamine

1-(2-methoxyethyl)-3,3-dimethylpiperazine

  • MDL number MFCD16301445
  • Cas Number : 1225500-78-6
  • Molecular Weight : 172.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-263548

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-263548
Availability details for
Quantity
  • Availability
  • Add to Cart