ENA497680355 Enamine

2-[(2-fluorophenyl)formamido]pentanedioic acid

  • MDL number MFCD12567370
  • Cas Number : 1234201-11-6
  • Molecular Weight : 269.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-263549

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-263549
Availability details for
Quantity
  • Availability
  • Add to Cart