ENA497680358 Enamine

3-phenyl-5,6-dihydro-1,4-oxathiine-2-carboxylic acid

  • MDL number MFCD09880367
  • Cas Number : 166270-89-9
  • Molecular Weight : 222.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-263956

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-263956
Availability details for
Quantity
  • Availability
  • Add to Cart