ENA497680361 Enamine

3-chloro-4,5-dimethoxyaniline

  • MDL number MFCD20542824
  • Cas Number : 90006-54-5
  • Molecular Weight : 187.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-263961

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-263961
Availability details for
Quantity
  • Availability
  • Add to Cart