ENA497680366 Enamine

(5-amino-1-benzyl-1H-1,2,3-triazol-4-yl)methanol

  • MDL number MFCD01109278
  • Cas Number : 49789-99-3
  • Molecular Weight : 204.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-263555

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-263555
Availability details for
Quantity
  • Availability
  • Add to Cart