ENA497680368 Enamine

dimethyl({[2-(5-methylfuran-2-yl)-1,3-thiazol-4-yl]methyl})amine

  • MDL number MFCD13546660
  • Cas Number : 1197527-61-9
  • Molecular Weight : 222.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-263558

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-263558
Availability details for
Quantity
  • Availability
  • Add to Cart