ENA497680371 Enamine

3-methyl-2-(thiophene-3-carbonyl)butanenitrile

  • MDL number MFCD14621508
  • Cas Number : 1250351-55-3
  • Molecular Weight : 193.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-264083

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-264083
Availability details for
Quantity
  • Availability
  • Add to Cart